3-Brom-5-fluor-2-methoxypyridin

Lieferant: ACTIVATE SCIENTIFIC

AS33975G1 AS33975G5
ACTIAS33975G1EA 0 CHF
ACTIAS33975G1 ACTIAS33975G5
3-Brom-5-fluor-2-methoxypyridin
3-Bromo-5-fluoro-2-methoxypyridine
Formel: C₆H₅BrFNO
MDL Number: MFCD04972397
CAS-Nummer: 884494-81-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR