Methyl-3-fluor-4-hydroxybenzoat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Methyl 3-fluoro-4-hydroxybenzoate

AS25002G25 AS25002G5
ACTIAS25002G25EA 160 CHF
ACTIAS25002G25 ACTIAS25002G5
Methyl-3-fluor-4-hydroxybenzoat
Methyl-3-fluor-4-hydroxybenzoat
Formel: C₈H₇FO₃
MW: 170,14 g/mol
MDL Number: MFCD00215838
CAS-Nummer: 403-01-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR