Methyl-4-brom-2-fluorbenzoat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Methyl 4-bromo-2-fluorobenzoate

AS22591G25 AS22591G5
ACTIAS22591G25EA 45 CHF
ACTIAS22591G25 ACTIAS22591G5
Methyl-4-brom-2-fluorbenzoat
Methyl-4-brom-2-fluorbenzoat
Formel: C₈H₆BrFO₂
MW: 233,04 g/mol
MDL Number: MFCD06203731
CAS-Nummer: 179232-29-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR