Methyl-2-brom-4-fluorbenzoat
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
Methyl 2-bromo-4-fluorobenzoate
AS23509G100
AS23509G25
AS23509G5
ACTIAS23509G100EA
366
CHF
ACTIAS23509G100
ACTIAS23509G25
ACTIAS23509G5
Methyl-2-brom-4-fluorbenzoat
Methyl-2-brom-4-fluorbenzoat
Formel:
C₈H₆BrFO₂ MW: 233,04 g/mol |
MDL Number:
MFCD06204026 CAS-Nummer: 653-92-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...