Yttrium(III)trifluormethansulfonat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Yttrium(III) trifluoromethanesulphonate, Yttrium triflate

AS23543G100 AS23543G25
ACTIAS23543G100EA 0 CHF
ACTIAS23543G100 ACTIAS23543G25
Yttrium(III)trifluormethansulfonat
Yttrium(III)trifluormethansulfonat
Formel: C₃F₉O₉S₃Y
MDL Number: MFCD00209623
CAS-Nummer: 52093-30-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR