Lanthan(III)trifluormethansulfonat

Lieferant: ACTIVATE SCIENTIFIC

AS23546G100 AS23546G25
ACTIAS23546G100EA 0 CHF
ACTIAS23546G100 ACTIAS23546G25
Lanthan(III)trifluormethansulfonat
Lanthan(III)trifluormethansulfonat
Formel: C₃F₉LaO₉S₃
MDL Number: MFCD00077560
CAS-Nummer: 52093-26-2
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR