1-(4-Aminophenyl)-4-methylpiperazin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-(4-Methylpiperazino)aniline, 4-(4-Methylpiperazin-1-yl)phenylamine, 4-(4-Methyl-1-piperazinyl)aniline

AS20589G100 AS20589G25 AS20589G5
ACTIAS20589G100EA 232 CHF
ACTIAS20589G100 ACTIAS20589G25 ACTIAS20589G5
1-(4-Aminophenyl)-4-methylpiperazin
1-(4-Aminophenyl)-4-methylpiperazin
Formel: C₁₁H₁₇N₃
MW: 191.28 g/mol
MDL Number: MFCD00172703
CAS-Nummer: 16153-81-4
UN: 3263
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR