3-Chlor-5-nitrobenzonitril

Lieferant: ACTIVATE SCIENTIFIC

AS21386G1 AS21386G5
ACTIAS21386G1EA 0 CHF
ACTIAS21386G1 ACTIAS21386G5
3-Chlor-5-nitrobenzonitril
3-Chlor-5-nitrobenzonitril
Formel: C₇H₃ClN₂O₂
MW: 182.57 g/mol
MDL Number: MFCD00463805
CAS-Nummer: 34662-30-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR