2-Chlor-4-nitrobenzonitril
Lieferant: ACTIVATE SCIENTIFIC
AS22669G100
AS22669G25
AS22669G5
ACTIAS22669G100EA
0
CHF
ACTIAS22669G100
ACTIAS22669G25
ACTIAS22669G5
2-Chlor-4-nitrobenzonitril
2-Chlor-4-nitrobenzonitril
Formel:
C₇H₃ClN₂O₂ |
MDL Number:
MFCD00052203 CAS-Nummer: 28163-00-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...