3-Fluor-5-nitrobenzonitril
Lieferant: ACTIVATE SCIENTIFIC
AS35699G1
AS35699G10
AS35699G5
ACTIAS35699G1EA
0
CHF
ACTIAS35699G1
ACTIAS35699G10
ACTIAS35699G5
3-Fluor-5-nitrobenzonitril
3-Fluor-5-nitrobenzonitril
Formel:
C₇H₃FN₂O₂ |
MDL Number:
MFCD05864356 CAS-Nummer: 110882-60-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...