2-Amino-5-iod-3-methylpyridin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 5-Iodo-3-methylpyridin-2-amine

AS33590G1 AS33590G5
ACTIAS33590G1EA 0 CHF
ACTIAS33590G1 ACTIAS33590G5
2-Amino-5-iod-3-methylpyridin
2-Amino-5-iodo-3-methylpyridine
Formel: C₆H₇IN₂
MW: 234.04 g/mol
MDL Number: MFCD02102422
CAS-Nummer: 166266-19-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR