3-Amino-2-fluor-6-methylpyridin

Lieferant: ACTIVATE SCIENTIFIC

AS7165G1 AS7165G5
ACTIAS7165G1EA 0 CHF
ACTIAS7165G1 ACTIAS7165G5
3-Amino-2-fluor-6-methylpyridin
3-Amino-2-fluoro-6-methylpyridine
Formel: C₆H₇FN₂
MW: 126.13 g/mol
MDL Number: MFCD03095221
CAS-Nummer: 374633-34-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR