1-Aminocyclopentancarbonsäure

Lieferant: ACTIVATE SCIENTIFIC
Achtung

AS20416G25 AS20416G5
ACTIAS20416G25EA 0 CHF
ACTIAS20416G25 ACTIAS20416G5
1-Aminocyclopentancarbonsäure
1-Aminocyclopentancarbonsäure
Formel: C₆H₁₁NO₂
Siedepunkt: 254 °C (1013 hPa)
Melting Pt: 163 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00001381
CAS-Nummer: 52-52-8
EINECS: 200-144-6
UN: 2811
ADR: 6.1,III
Merck Index: 13,02763

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR