8-Hydroxychinaldin

Lieferant: ACTIVATE SCIENTIFIC
Achtung

AS33511G10 AS33511G100
ACTIAS33511G10EA 0 CHF
ACTIAS33511G10 ACTIAS33511G100
8-Hydroxychinaldin
8-Hydroxychinaldin
Formel: C₁₀H₉NO
Siedepunkt: 267 °C (1013 hPa)
Melting Pt: 70…72 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00006765
CAS-Nummer: 826-81-3
EINECS: 212-562-6
UN: 3077
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR