8-Chinolinsulfonsäure

Lieferant: ACTIVATE SCIENTIFIC
Achtung

Synonyms: 8-Quinolinesulphonic acid

AS21609G100 AS21609G25
ACTIAS21609G100EA 0 CHF
ACTIAS21609G100 ACTIAS21609G25
8-Chinolinsulfonsäure
8-Chinolinsulfonsäure
Formel: C₉H₇NO₃S
MW: 209.23 g/mol
Melting Pt: > 300 °C
MDL Number: MFCD00024037
CAS-Nummer: 85-48-3
EINECS: 201-611-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR