5-Nitrochinolin

Lieferant: ACTIVATE SCIENTIFIC

AS20718G100 AS20718G25
ACTIAS20718G100EA 0 CHF
ACTIAS20718G100 ACTIAS20718G25
5-Nitrochinolin
5-Nitroquinoline
Formel: C₉H₆N₂O₂
MW: 174.16 g/mol
Siedepunkt: 323
Melting Pt: 71
MDL Number: MFCD00006790
CAS-Nummer: 607-34-1
EINECS: 210-134-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR