3-Aminochinolin

Lieferant: ACTIVATE SCIENTIFIC
Achtung

Synonyms: Quinolin-3-amine, Quinolin-3-ylamine

AS40308G1 AS40308G5
ACTIAS40308G1EA 0 CHF
ACTIAS40308G1 ACTIAS40308G5
3-Aminochinolin
3-Aminoquinoline
Formel: C₉H₈N₂
MW: 144,18 g/mol
Siedepunkt: 311 °C (1013 hPa)
Melting Pt: 84 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00006772
CAS-Nummer: 580-17-6
EINECS: 209-455-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR