2-Aminochinolin
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
Quinolin-2-ylamine
AS1684G1
AS1684G5
ACTIAS1684G1EA
0
CHF
ACTIAS1684G1
ACTIAS1684G5
2-Aminochinolin
2-Aminoquinoline
Formel:
C₉H₈N₂ MW: 144,18 g/mol Siedepunkt: 305 °C (1013 hPa) Melting Pt: 130 °C |
MDL Number:
MFCD00038079 CAS-Nummer: 580-22-3 EINECS: 209-458-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...