5-Chlorchinolin
Lieferant: ACTIVATE SCIENTIFIC
|
Achtung
|
AS35427G1
AS35427G5
ACTIAS35427G1EA
0
CHF
ACTIAS35427G1
ACTIAS35427G5
5-Chlorchinolin
5-Chloroquinoline
Formel:
C₉H₆ClN MW: 163,61 g/mol Lager Temperatur: Kühlschrank |
MDL Number:
MFCD00239413 CAS-Nummer: 635-27-8 |
Learn more
![](/m-ch.vwr.com/de_CH/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...