8-Chinolincarbonsäure
Lieferant: ACTIVATE SCIENTIFIC
|
Achtung
|
Synonyms:
Quinoline-8-carboxylic acid
AS1358G1
AS1358G5
ACTIAS1358G1EA
0
CHF
ACTIAS1358G1
ACTIAS1358G5
8-Chinolincarbonsäure
8-Chinolincarbonsäure
Formel:
C₁₀H₇NO₂ MW: 173,17 g/mol |
MDL Number:
MFCD00047619 CAS-Nummer: 86-59-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...