6-Chinolincarbonsäure

Lieferant: ACTIVATE SCIENTIFIC GMBH
Achtung

ACTIAS7015G100EA 0 CHF
ACTIAS7015G100 ACTIAS7015G10 ACTIAS7015G25
6-Chinolincarbonsäure
6-Chinolincarbonsäure
Formel: C₁₀H₇NO₂
MW: 173,17 g/mol
Siedepunkt: 345 °C (1013 hPa)
Melting Pt: 294…295 °C
MDL Number: MFCD00047613
CAS-Nummer: 10349-57-2
EINECS: 233-761-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR