4-Chinolincarbonsäure
Lieferant: ACTIVATE SCIENTIFIC
|
Achtung
|
Synonyms:
Quinoline-4-carboxylic acid, NSC 13138
AS7053G1
AS7053G5
ACTIAS7053G1EA
0
CHF
ACTIAS7053G1
ACTIAS7053G5
4-Chinolincarbonsäure
4-Chinolincarbonsäure
Formel:
C₁₀H₇NO₂ MW: 173,17 g/mol Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00006782 CAS-Nummer: 486-74-8 EINECS: 207-640-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...