8-Chinolincarbaldehyd

Lieferant: ACTIVATE SCIENTIFIC
Achtung

Synonyms: Quinoline-8-carbaldehyde, 8-Formylquinoline, 8-Quinolinecarboxaldehyde, Quinoline-8-carboxaldehyde

AS1357G1 AS1357G5
ACTIAS1357G1EA 0 CHF
ACTIAS1357G1 ACTIAS1357G5
8-Chinolincarbaldehyd
8-Chinolincarbaldehyd
Formel: C₁₀H₇NO
MW: 157,17 g/mol
MDL Number: MFCD00805837
CAS-Nummer: 38707-70-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR