5-Chinolincarbaldehyd

Lieferant: ACTIVATE SCIENTIFIC
Achtung

Synonyms: Quinoline-5-carboxaldehyde, Quinoline-5-carbaldehyde

AS20191G1 AS20191G5
ACTIAS20191G1EA 196 CHF
ACTIAS20191G1 ACTIAS20191G5
5-Chinolincarbaldehyd
5-Chinolincarbaldehyd
Formel: C₁₀H₇NO
MW: 157,17 g/mol
MDL Number: MFCD00805835
CAS-Nummer: 22934-41-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR