8-Bromchinolin
Lieferant: ACTIVATE SCIENTIFIC GMBH
|
Achtung
|
ACTIAS7098G5EA
0
CHF
ACTIAS7098G5
ACTIAS7098G25
8-Bromchinolin
8-Bromoquinoline
Formel:
C₉H₆BrN MW: 208,06 g/mol |
MDL Number:
MFCD00191859 CAS-Nummer: 16567-18-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...