2-Bromchinolin

Lieferant: ACTIVATE SCIENTIFIC
Achtung

AS1087G1 AS1087G5
ACTIAS1087G1EA 0 CHF
ACTIAS1087G1 ACTIAS1087G5
2-Bromchinolin
2-Bromoquinoline
Formel: C₉H₆BrN
MW: 208,06 g/mol
MDL Number: MFCD00234480
CAS-Nummer: 2005-43-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR