3,5-Dimethylbenzonitril

Lieferant: ACTIVATE SCIENTIFIC

AS24890G100 AS24890G25
ACTIAS24890G100EA 240 CHF
ACTIAS24890G100 ACTIAS24890G25
3,5-Dimethylbenzonitril
3,5-Dimethylbenzonitril
Formel: C₉H₉N
MW: 131.18 g/mol
MDL Number: MFCD00060670
CAS-Nummer: 22445-42-7
EINECS: 245-001-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR