4-Methyl-2-thiophenboronsäure
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
(4-Methylthiophen-2-yl)boronic acid, 4-Methylthiophene-2-boronic acid
AS50890G1
AS50890G5
ACTIAS50890G1EA
0
CHF
ACTIAS50890G1
ACTIAS50890G5
4-Methyl-2-thiophenboronsäure
4-Methyl-2-thiopheneboronic acid
Formel:
C₅H₇BO₂S MW: 141,99 g/mol |
MDL Number:
MFCD01319040 CAS-Nummer: 162607-15-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...