5-Brom-2-thiophencarbaldehyd
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
5-Bromo-2-thiophenecarboxaldehyde
AS33822G10
AS33822G100
ACTIAS33822G10EA
0
CHF
ACTIAS33822G10
ACTIAS33822G100
5-Brom-2-thiophencarbaldehyd
5-Brom-2-thiophencarbaldehyd
Formel:
C₅H₃BrOS MW: 191,05 g/mol |
MDL Number:
MFCD00005432 CAS-Nummer: 4701-17-1 EINECS: 225-176-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...