5-Formyl-2-thiophenboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 5-Formylthiophen-2-boronic acid, 5-Formylthiophene-2-boronic acid

AS50731G1 AS50731G5
ACTIAS50731G1EA 0 CHF
ACTIAS50731G1 ACTIAS50731G5
5-Formyl-2-thiophenboronsäure
5-Formyl-2-thiophenboronsäure
Formel: C₅H₅BO₃S
MW: 155,97 g/mol
MDL Number: MFCD02093666
CAS-Nummer: 4347-33-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR