3-(Methylthio)phenylboronsäure
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
3-Thioanisoleboronic acid, 3-(Methylthio)benzeneboronic acid
AS50892G100
AS50892G5
ACTIAS50892G100EA
0
CHF
ACTIAS50892G100
ACTIAS50892G5
3-(Methylthio)phenylboronsäure
3-(Methylthio)phenylboronsäure
Formel:
C₇H₉BO₂S MW: 168,02 g/mol |
MDL Number:
MFCD01319105 CAS-Nummer: 128312-11-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...