2-(Methylthio)phenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: (2-(Methylthio)phenyl)boronic acid, 2-(Methylthio)benzeneboronic acid

AS50891G1 AS50891G5
ACTIAS50891G1EA 0 CHF
ACTIAS50891G1 ACTIAS50891G5
2-(Methylthio)phenylboronsäure
2-(Methylthio)phenylboronsäure
Formel: C₇H₉BO₂S
MW: 168.02 g/mol
MDL Number: MFCD01318165
CAS-Nummer: 168618-42-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR