(±)-4-Brom-α-methylbenzylamin
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
4-Bromo-α-methylbenzylamine, (+/-)-1-(4-Bromophenyl)ethylamine, (±)-4-Bromo-α-methylbenzylamine
AS33218G10
AS33218G5
ACTIAS33218G10EA
0
CHF
ACTIAS33218G10
ACTIAS33218G5
(±)-4-Brom-α-methylbenzylamin
(±)-4-Brom-α-methylbenzylamin
Formel:
C₈H₁₀BrN |
MDL Number:
MFCD00025548 CAS-Nummer: 24358-62-1 EINECS: 246-200-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...