(S)-(-)-4-Methoxy-α-methylbenzylamin
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
(S)-1-(4-Methoxyphenyl)ethylamine, (S)-(-)-1-(4-Methoxyphenyl)ethylamine
AS34126G10
AS34126G100
ACTIAS34126G10EA
0
CHF
ACTIAS34126G10
ACTIAS34126G100
(S)-(-)-4-Methoxy-α-methylbenzylamin
(S)-(-)-4-Methoxy-α-methylbenzylamin
Formel:
C₉H₁₃NO MW: 151,21 g/mol |
MDL Number:
MFCD00671660 CAS-Nummer: 41851-59-6 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...