(R)-(+)-4-Methoxy-α-methylbenzylamin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: (R)-1-(4-Methoxyphenyl)ethan-1-amine, (R)-(+)-1-(4-Methoxyphenyl)ethylamine

AS32317G5 AS32317G10
ACTIAS32317G5EA 0 CHF
ACTIAS32317G5 ACTIAS32317G10
(R)-(+)-4-Methoxy-α-methylbenzylamin
(R)-(+)-4-Methoxy-α-methylbenzylamin
Formel: C₉H₁₃NO
MW: 151.21 g/mol
MDL Number: MFCD00671659
CAS-Nummer: 22038-86-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR