4-Aminomorpholin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: N-Aminomorpholine

AS20156G100 AS20156G25 AS20156G5
ACTIAS20156G100EA 0 CHF
ACTIAS20156G100 ACTIAS20156G25 ACTIAS20156G5
4-Aminomorpholin
4-Aminomorpholin
Formel: C₄H₁₀N₂O
MDL Number: MFCD00006174
CAS-Nummer: 4319-49-7
EINECS: 224-347-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR