2-Chlorphenethylbromid
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
1-(2-Bromoethyl)-2-chlorobenzene, 2-Chlorophenethyl Bromide, 2-(2-Chlorophenyl)ethyl bromide, 2-Chlorophenethyl bromide
AS21533G25
AS21533G5
ACTIAS21533G25EA
0
CHF
ACTIAS21533G25
ACTIAS21533G5
2-Chlorphenethylbromid
2-Chlorphenethylbromid
Formel:
C₈H₈BrCl |
MDL Number:
MFCD04117464 CAS-Nummer: 16793-91-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...