1,3-Diethylharnstoff

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: N,N'-Diethylurea

AS20170G100 AS20170G25
ACTIAS20170G100EA 105 CHF
ACTIAS20170G100 ACTIAS20170G25
1,3-Diethylharnstoff
1,3-Diethylharnstoff
Formel: C₅H₁₂N₂O
MW: 116.16 g/mol
MDL Number: MFCD00009028
CAS-Nummer: 623-76-7
EINECS: 210-811-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR