3,5-Dimethylbenzaldehyd

Lieferant: ACTIVATE SCIENTIFIC

AS33223G1 AS33223G10 AS33223G5
ACTIAS33223G1EA 0 CHF
ACTIAS33223G1 ACTIAS33223G10 ACTIAS33223G5
3,5-Dimethylbenzaldehyd
3,5-Dimethylbenzaldehyde
Formel: C₉H₁₀O
MW: 134,18 g/mol
Dichte: 1.09 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00082777
CAS-Nummer: 5779-95-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR