3,4-Dimethylbenzaldehyd
Lieferant: ACTIVATE SCIENTIFIC
AS21516G100
AS21516G25
ACTIAS21516G100EA
0
CHF
ACTIAS21516G100
ACTIAS21516G25
3,4-Dimethylbenzaldehyd
3,4-Dimethylbenzaldehyde
Formel:
C₉H₁₀O MW: 134.18 g/mol Siedepunkt: 68 °C (0.8 mmHg) Dichte: 1.012 g/cm³ (20 °C) |
MDL Number:
MFCD00016612 CAS-Nummer: 5973-71-7 EINECS: 227-770-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...