α'-Brom-α,α,α-trifluor-o-xylol

Lieferant: ACTIVATE SCIENTIFIC
Gefahr

Synonyms: 2-(Trifluoromethyl)benzyl bromide, 1-(Bromomethyl)-2-(trifluoromethyl)benzene, 2-(Bromomethyl)benzotrifluoride

AS24100G100 AS24100G25
ACTIAS24100G100EA 0 CHF
ACTIAS24100G100 ACTIAS24100G25
α'-Brom-α,α,α-trifluor-o-xylol
α'-Brom-α,α,α-trifluor-o-xylol
Formel: C₈H₆BrF₃
Siedepunkt: 116…118 °C (60 mmHg)
Melting Pt: 30 °C
Dichte: 1.57 g/cm³ (20 °C)
MDL Number: MFCD00013558
CAS-Nummer: 395-44-8
EINECS: 206-901-7
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR