4-Isopropylanilin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: p-Cumidine, 4-Aminocumene

AS21354G100 AS21354G25
ACTIAS21354G100EA 0 CHF
ACTIAS21354G100 ACTIAS21354G25
4-Isopropylanilin
4-Isopropylanilin
Formel: C₉H₁₃N
MW: 135.21 g/mol
Siedepunkt: 226
Melting Pt: –63 °C
Dichte: 0.9526 g/cm³ (20 °C)
Flash-Pt: 92
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00007900
CAS-Nummer: 99-88-7
EINECS: 202-797-2
UN: 1760
ADR: 8 III
Merck Index: 13,02647

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR