3-tert-Butylanilin

Lieferant: ACTIVATE SCIENTIFIC
Achtung

AS32656G1 AS32656G5
ACTIAS32656G1EA 0 CHF
ACTIAS32656G1 ACTIAS32656G5
3-tert-Butylanilin
3-tert-Butylanilin
Formel: C₁₀H₁₅N
MDL Number: MFCD00125078
CAS-Nummer: 5369-19-7
EINECS: 226-361-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR