2,5-Difluortoluol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 1,4-Difluoro-2-methylbenzene

AS24911G100 AS24911G25
ACTIAS24911G100EA 0 CHF
ACTIAS24911G100 ACTIAS24911G25
2,5-Difluortoluol
2,5-Difluortoluol
Formel: C₇H₆F₂
Siedepunkt: 117 °C (1013 hPa)
Melting Pt: –35 °C
Dichte: 1.36 g/cm³ (20 °C)
MDL Number: MFCD00000347
CAS-Nummer: 452-67-5
EINECS: 207-205-6
UN: 2388
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR