2-Chlor-3-nitrotoluol
Lieferant: ACTIVATE SCIENTIFIC
AS32982G1
AS32982G5
ACTIAS32982G1EA
0
CHF
ACTIAS32982G1
ACTIAS32982G5
2-Chlor-3-nitrotoluol
2-Chlor-3-nitrotoluol
Formel:
C₇H₆ClNO₂ |
MDL Number:
MFCD00134653 CAS-Nummer: 3970-40-9 EINECS: 223-591-9 UN: 2433 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...