(4-Chlorphenyl)aceton

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-Chlorophenylacetone

AS24978G100 AS24978G25
ACTIAS24978G100EA 0 CHF
ACTIAS24978G100 ACTIAS24978G25
(4-Chlorphenyl)aceton
(4-Chlorphenyl)aceton
Formel: C₉H₉ClO
MW: 168,62 g/mol
MDL Number: MFCD00045214
CAS-Nummer: 5586-88-9
EINECS: 226-986-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR