6-Nitro-m-toluylsäure

Lieferant: ACTIVATE SCIENTIFIC

AS21842G100 AS21842G25
ACTIAS21842G100EA 0 CHF
ACTIAS21842G100 ACTIAS21842G25
6-Nitro-m-toluylsäure
6-Nitro-m-toluylsäure
Formel: C₈H₇NO₄
Siedepunkt: 364 °C (1013 hPa)
Melting Pt: 137…138 °C
MDL Number: MFCD00007368
CAS-Nummer: 3113-72-2
EINECS: 221-481-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR