4-Methoxy-3-methylbenzaldehyd
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
3-Methyl-4-anisaldehyde
AS32739G1
AS32739G10
AS32739G5
ACTIAS32739G1EA
0
CHF
ACTIAS32739G1
ACTIAS32739G10
ACTIAS32739G5
4-Methoxy-3-methylbenzaldehyd
4-Methoxy-3-methylbenzaldehyd
Formel:
C₉H₁₀O₂ MW: 150,18 g/mol Siedepunkt: 80 °C (1013 hPa) Dichte: 1.082 g/cm³ (16 °C) Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00003375 CAS-Nummer: 32723-67-4 EINECS: 251-177-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...