4-tert-Butylbenzaldehyd

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-(tert-Butyl)benzaldehyde

AS24410G100 AS24410G25
ACTIAS24410G100EA 0 CHF
ACTIAS24410G100 ACTIAS24410G25
4-tert-Butylbenzaldehyd
4-tert-Butylbenzaldehyd
Formel: C₁₁H₁₄O
MW: 162,23 g/mol
Siedepunkt: 58…60 °C (0.2 mmHg)
Dichte: 0.971 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00035742
CAS-Nummer: 939-97-9
EINECS: 213-367-9
UN: 3082
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR