4-Brom-2-nitrotoluol

Lieferant: ACTIVATE SCIENTIFIC
Achtung

AS21187G100 AS21187G25
ACTIAS21187G100EA 0 CHF
ACTIAS21187G100 ACTIAS21187G25
4-Brom-2-nitrotoluol
4-Brom-2-nitrotoluol
Formel: C₇H₆BrNO₂
MW: 216,03 g/mol
Siedepunkt: 75…82 °C (0,3 mmHg)
Melting Pt: 43…46 °C
MDL Number: MFCD00041243
CAS-Nummer: 60956-26-5
EINECS: 262-536-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR